Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse PLC-delta3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5315
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5315 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5315 Supplier R&D Systems Supplier No. AF5315
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

PLC-delta 3 Polyclonal specifically detects PLC-delta 3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen PLC-delta 3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q8N3E9
Gene Alias EC 3.1.4.11, KIAA1964, MGC71172, Phosphoinositide phospholipase C-delta-3, phospholipase C, delta 3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3, Phospholipase C-delta-3, PLC delta3, PLCD3, PLCdelta 3, PLC-delta-3
Gene Symbols PLCD3
Host Species Goat
Immunogen E. coli-derived recombinant human PLC-delta 3 Lys148-Ala344 Accession # Q8N3E9
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113026
Test Specificity Detects human and mouse PLC-delta 3 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.