Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse PLC-gamma 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3716SP
This item is not returnable.
View return policy
Description
PLC-gamma 2 Monoclonal specifically detects PLC-gamma 2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
PLC-gamma 2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P16885 | |
PLCG2 | |
E. coli-derived recombinant human PLC-gamma 2 Pro400-Tyr600 Accession # P16885 | |
25 μg | |
Primary | |
Detects human and mouse PLC-gamma 2. | |
Human, Mouse | |
Purified |
Western Blot, Immunocytochemistry | |
346404 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.1.4.11, Phosphoinositide phospholipase C-gamma-2, phospholipase C gamma 2, phospholipase C, gamma 2 (phosphatidylinositol-specific), phospholipase C, gamma 2 (phosphatidylyinositol-specific), Phospholipase C-gamma-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2, Phospholipase C-IV, PLCG2, PLCgamma 2, PLC-gamma-2, PLC-IV | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5336 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction