Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Plexin A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5486SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5486SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5486SP Supplier R&D Systems Supplier No. MAB5486SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Plexin A2 Monoclonal specifically detects Plexin A2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Plexin A2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 583603
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P70207
Gene Alias FLJ11751, FLJ30634, KIAA0463plexin 2, OCT, OCTSemaphorin receptor OCT, plexin A2, PLXN2, PLXN2plexin-A2, PLXNA2, transmembrane protein OCT
Gene Symbols PLXNA2
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin A2 Met35-Pro1237 Accession # P70207
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5362
Test Specificity Detects mouse Plexin A2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Plexin A4, B1, B2, D1, recombinant mouse Plexin A1, A3, B3, or C1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.