Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Plexin A2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB5486
This item is not returnable.
View return policy
Description
Plexin A2 Monoclonal specifically detects Plexin A2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Plexin A2 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P70207 | |
PLXNA2 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin A2 Met35-Pro1237 Accession # P70207 | |
100 μg | |
Primary | |
Detects mouse Plexin A2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Plexin A4, B1, B2, D1, recombinant mouse Plexin A1, A3, B3, or C1 is observed. | |
Human, Mouse | |
Purified |
Western Blot, Immunocytochemistry | |
583603 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
FLJ11751, FLJ30634, KIAA0463plexin 2, OCT, OCTSemaphorin receptor OCT, plexin A2, PLXN2, PLXN2plexin-A2, PLXNA2, transmembrane protein OCT | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5362 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction