Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Plexin C1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB3887
This item is not returnable.
View return policy
Description
Plexin C1 Monoclonal specifically detects Plexin C1 in Human, Mouse samples. It is validated for Immunohistochemistry.Specifications
Plexin C1 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
O60486 | |
PLXNC1 | |
E. coli-derived recombinant human Plexin C1 Ile49-Ser161 Accession # O60486 | |
100 μg | |
Primary | |
Detects human Plexin C1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Plexin A4, B1, B2, B3, D1, recombinant mouse Plexin A1, A2, or A3 is observed. | |
Human, Mouse | |
Purified |
Immunohistochemistry | |
544232 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD232, CD232 antigen, plexin (semaphorin receptor), plexin C1, PLXNC1, PLXN-C1, receptor for viral semaphorin protein, receptor for virally-encoded semaphorin, VESPR, VESPRplexin-C1, Virus-encoded semaphorin protein receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10154 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction