Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Polypeptide GalNAc Transferase 10/GALNT10 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7575
This item is not returnable.
View return policy
Description
Polypeptide GalNac Transferase 10/GALNT10 Polyclonal specifically detects Polypeptide GalNac Transferase 10/GALNT10 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Polypeptide GalNac Transferase 10/GALNT10 | |
Polyclonal | |
Unconjugated | |
Q86SR1 | |
GALNT10 | |
Mouse myeloma cell line NS0-derived recombinant human Polypeptide GalNac Transferase 10/GALNT10 Leu71-Asn603 Accession # Q86SR1 | |
100 μg | |
Primary | |
Detects human and mouse Polypeptide GalNac Transferase 10/GALNT10 in Western blots. In direct ELISA, less than 1% cross-reactivity with recombinant human (rh) GALNT1 and rhGALNT4 is observed. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
DKFZp586H0623, EC 2.4.1.41, FLJ00205, FLJ11715, GalNAc transferase 10, GalNAc-T10, GALNACT10, Polypeptide GalNAc transferase 10, polypeptide N-acetylgalactosaminyltransferase 10, PPGALNACT10, pp-GaNTase 10, PPGANTASE10, Protein-UDP acetylgalactosaminyltransferase 10, UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 10, UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 10 (GalNAc-T10) | |
Sheep | |
Affinity Purified | |
RUO | |
55568 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction