Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse PRDM16/MEL1 Antibody, R&D Systems™

Catalog No. AF6295
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6295 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6295 Supplier R&D Systems Supplier No. AF6295
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 12 publications

PRDM16/MEL1 Polyclonal specifically detects PRDM16/MEL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen PRDM16/MEL1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. A2A935.1
Gene Alias MDS1/EVI1-like gene 1, MEL1, MEL1KIAA1675PFM13MGC166915, PFM13, PR domain containing 16, PR domain zinc finger protein 16, PR domain-containing protein 16, Transcription factor MEL1
Gene Symbols PRDM16
Host Species Sheep
Immunogen E. coli-derived recombinant mouse PRDM16 Lys537-Glu688 Accession # A2A935.1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63976
Test Specificity Detects mouse and human PRDM16 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.