Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Primate Dopamine beta-Hydroxylase C-Terminus Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems PPS066
This item is not returnable.
View return policy
Description
Dopamine beta-Hydroxylase Polyclonal specifically detects Dopamine beta-Hydroxylase in Human, Mouse, Primate samples. It is validated for Western Blot.Specifications
Dopamine beta-Hydroxylase | |
Polyclonal | |
Western Blot 1:1000 dilution | |
DBH | |
Dopamine beta-Hydroxylase | |
100 μL | |
Primary | |
Human, monkey, bovine, and rabbit ∼75 kDa Dopamine beta-hydroxylase (D beta H) | |
For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
Western Blot | |
Unconjugated | |
DBH, DBM, dopamine beta-hydroxylase, Dopamine betaHydroxylase, dopamine beta-hydroxylase (dopamine beta-monooxygenase), Dopamine beta-monooxygenase, DOPBHY, EC 1.14.17.1 | |
Sheep | |
Affinity Purified | |
RUO | |
1621 | |
Human, Mouse, Primate | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction