Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Primate SLC6A3/DAT1 C-Terminus Antibody, R&D Systems™

Catalog No. PPS068
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPS068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPS068 Supplier R&D Systems Supplier No. PPS068
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SLC6A3/DAT1 Polyclonal specifically detects SLC6A3/DAT1 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen SLC6A3/DAT1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000 dilution, Immunohistochemistry 1:1000 dilution
Gene Alias DA transporter, DAT1, DATDAT1Solute carrier family 6 member 3, SLC6A3, sodium-dependent dopamine transporter, solute carrier family 6 (neurotransmitter transporter, dopamine), member 3, VNTR
Gene Symbols SLC6A3
Host Species Rabbit
Immunogen SLC6A3/DAT1
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6531
Test Specificity Specific for the ∼88 kDa DAT protein in Western blots of SDS-solubilized human and mouse striatal samples and in IHC applications with formaldehyde-fixed human and monkey (Macaque) brain sections.
Target Species Human, Mouse, Primate
Content And Storage For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.