Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Proinsulin Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC13361G100UG
This item is not returnable.
View return policy
Description
Proinsulin Monoclonal specifically detects Proinsulin in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| Proinsulin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P01308 | |
| INS | |
| E. coli-derived recombinant human Proinsulin Phe25-Asn110 Accession # P01308 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Proinsulin. Does not detect mature insulin. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 253627 | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3630 | |
| Human, Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction