Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Proinsulin Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM13361
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM13361 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM13361 Supplier R&D Systems Supplier No. BAM13361
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Proinsulin Monoclonal specifically detects Proinsulin in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Proinsulin
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 253627
Concentration LYOPH
Conjugate Biotin
Dilution Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. P01308
Gene Alias IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Gene Symbols INS
Host Species Mouse
Immunogen E. coli-derived recombinant human Proinsulin Phe25-Asn110 Accession # P01308
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity Detects human and mouse Proinsulin in direct ELISAs. In direct ELISAs, no cross-reactivity with mature Insulin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.