Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Proprotein Convertase 2/PCSK2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6018
This item is not returnable.
View return policy
Description
Proprotein Convertase 2/PCSK2 Polyclonal specifically detects Proprotein Convertase 2/PCSK2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.Specifications
Proprotein Convertase 2/PCSK2 | |
Polyclonal | |
Unconjugated | |
P16519 | |
PCSK2 | |
Chinese hamster ovary cell line CHO-derived recombinant human Proprotein Convertase 2/PCSK2 Gly110-Asn638 Accession # P16519 | |
100 μg | |
Primary | |
Detects human Proprotein Convertase 2/PCSK2 in direct ELISAs and, human and mouse Proprotein Convertase 2/PCSK2 in Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
EC 3.4.21, EC 3.4.21.94, KEX2-like endoprotease 2, NEC 2, NEC2, NEC2SPC2, neuroendocrine convertase 2, PC2, PC2NEC-2, PCSK2, Prohormone convertase 2, Proprotein convertase 2, proprotein convertase subtilisin/kexin type 2, SPC2 | |
Sheep | |
Affinity Purified | |
RUO | |
5126 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction