Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse PTEN Antibody, R&D Systems™

Mouse Monoclonal Antibody
$227.50 - $538.00
Specifications
| Antigen | PTEN |
|---|---|
| Clone | 217702R |
| Dilution | Western Blot 2 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, Knockout Validated 2 ug/mL |
| Applications | Western Blot, Flow Cytometry, KnockDown |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB847RSP
![]() |
R&D Systems
MAB847RSP |
25 μg |
Each for $227.50
|
|
|||||
MAB847R100
![]() |
R&D Systems
MAB847R100 |
100 μg |
Each for $538.00
|
|
|||||
Description
PTEN Monoclonal antibody specifically detects PTEN in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, KnockDownSpecifications
| PTEN | |
| Western Blot 2 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, Knockout Validated 2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BZS, EC 3.1.3.16,10q23del, EC 3.1.3.48, EC 3.1.3.67, GLM2, MMAC1, MMAC1 phosphatase and tensin homolog deleted on chromosome 10, MMAC1MGC11227, Mutated in multiple advanced cancers 1, phosphatase and tensin homologDEC, phosphatidylinositol-34,5-trisphosphate 3-phosphatase and dual-specificityprotein phosphatase PTEN, PTEN1, TEP1MHAM | |
| E. coli-derived recombinant human PTEN, Thr2-Val403, Accession # P60484 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 217702R | |
| Western Blot, Flow Cytometry, KnockDown | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P60484 | |
| 5728 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title