Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse PUM1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7628SP
This item is not returnable.
View return policy
Description
PUM1 Polyclonal specifically detects PUM1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
PUM1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HSPUM, KIAA0099PUML1, PUMH, PUMH1, PUMH1pumilio homolog 1, pumilio (Drosophila) homolog 1, pumilio homolog 1 (Drosophila), Pumilio-1, PUML1 | |
Sheep | |
Affinity Purified | |
RUO | |
9698 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Q14671 | |
PUM1 | |
E. coli-derived recombinant human PUM1 Pro230-Ala372 Accession # Q14671 | |
25 μg | |
Primary | |
Detects human and mouse PUM1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human PUM2 is observed. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction