Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Rab7a Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7789
This item is not returnable.
View return policy
Description
Rab7a Polyclonal specifically detects Rab7a in Human, Mouse samples. It is validated for Western Blot.Specifications
Rab7a | |
Polyclonal | |
Unconjugated | |
P51149 | |
RAB7A | |
E. coli-derived recombinant human Rab7a Thr2-Ser204 Accession # P51149 | |
100 μg | |
Primary | |
Detects human and mouse Rab7a in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Rab9a and rhRab9b is observed. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
CMT2B, FLJ20819, member RAS oncogene family, PRO2706, PSN, RAB7, RAB7A, member RAS oncogene family, Ras-associated protein RAB7, ras-related protein Rab-7a | |
Sheep | |
Affinity Purified | |
RUO | |
7879 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction