Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse RASSF2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5639SP
This item is not returnable.
View return policy
Description
RASSF2 Polyclonal specifically detects RASSF2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
RASSF2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
KIAA0168DKFZp781O1747, Ras association (RalGDS/AF-6) domain family 2, Ras association (RalGDS/AF-6) domain family member 2, ras association domain-containing protein 2, RASFADIN | |
Goat | |
Affinity Purified | |
RUO | |
9770 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
P50749 | |
RASSF2 | |
E. coli-derived recombinant human RASSF2 Lys100-Gly230 Accession # P50749 | |
25 μg | |
Primary | |
Detects endogenous human and mouse RASSF2 in Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction