Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat 14-3-3 eta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4420
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4420 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4420 Supplier R&D Systems Supplier No. AF4420
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

14-3-3 eta Polyclonal specifically detects 14-3-3 eta in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen 14-3-3 eta
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q04917
Gene Alias 1433 eta, 14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA1, YWHA114-3-3 eta, YWHAH
Gene Symbols YWHAH
Host Species Goat
Immunogen E. coli-derived recombinant human 14-3-3 eta Met1-Asn246 Accession # Q04917
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7533
Test Specificity Detects human, mouse, and rat 14-3-3 eta in Western blots. In Western Blot less than 1% cross reactivity with other recombinant human 14-3-3 isoforms.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.