Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat 14-3-3 sigma Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4424
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4424 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4424 Supplier R&D Systems Supplier No. AF4424
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

14-3-3 sigma/Stratifin Polyclonal specifically detects 14-3-3 sigma/Stratifin in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.

Specifications

Antigen 14-3-3 sigma/Stratifin
Applications Western Blot, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Knockout Validated
Gene Accession No. P31947
Gene Alias 14-3-3 protein sigma, 1433 sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, SFN, stratifin, YWHAS
Gene Symbols SFN
Host Species Goat
Immunogen E. coli-derived recombinant human 14-3-3 sigma Met1-Ser248 Accession # P31947
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2810
Test Specificity Detects human, mouse, and rat 14-3-3 sigma in Western blots. Does not cross react with other recombinant human 14-3-3 isoforms.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.