Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Akt1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1775
This item is not returnable.
View return policy
Description
AKT1 Monoclonal specifically detects AKT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
AKT1 | |
Monoclonal | |
LYOPH | |
Western Blot 0.2 ug/mL | |
P31749 | |
AKT1 | |
E. coli-derived recombinant human Akt1 Ser2-Ala480 Accession # P31749 | |
100 μg | |
Primary | |
Detects human, mouse, and rat Akt1 in Western blots. In Western blots, no cross-reactivity with recombinant human Akt2 or Akt3 is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
232790 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
207 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction