Learn More
Human/Mouse/Rat Akt1 Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Polyclonal Antibody
Supplier:  R&D Systems AF1775
Catalog No. AF1775
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
AKT1 Polyclonal specifically detects AKT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
Specifications
AKT1
Polyclonal
Unconjugated
P31749
AKT1
E. coli-derived recombinant human Akt1 Ser2-Ala480 Accession # P31749
50 μg
Primary
Detects human, mouse, and rat Akt1 in Western blots. Does not detect recombinant Akt2 or Akt3 in Western blots.
Human, Mouse, Rat
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL
AKT, AKT serine/threonine kinase 1, AKT1m, CWS6, EC 2.7.11, EC 2.7.11.1, PKB, PKB alpha, PKBMGC99656, PRKBA, Protein kinase B, Protein Kinase B Alpha, Proto-oncogene c-Akt, RAC, rac protein kinase alpha, RAC-ALPHA, RAC-alpha serine/threonine-protein kinas
Rabbit
Affinity Purified
RUO
207
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.