Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat alpha-Actinin 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8279SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8279SP 25 μg
MAB8279 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8279SP Supplier R&D Systems Supplier No. MAB8279SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha-Actinin 1 Monoclonal specifically detects alpha-Actinin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen alpha-Actinin 1
Applications Western Blot
Classification Monoclonal
Clone 910301
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P12814
Gene Alias actinin 1 smooth muscle, actinin, alpha 1, ACTN1, alphaActinin 1, alpha-Actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, BDPLT15, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1
Gene Symbols ACTN1
Host Species Mouse
Immunogen E. coli-derived recombinant human alpha-Actinin 1 Asn272-Thr500 Accession # P12814
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 87
Test Specificity Detects human alpha-Actinin 1 in ELISA, and human, mouse and rat alpha-Actinin 1 in Western Blot.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.