Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat alpha-Cardiac Actin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB93081SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB93081SP 25 μg
MAB93081100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB93081SP Supplier R&D Systems Supplier No. MAB93081SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Actin Alpha 1 Cardiac Muscle Monoclonal specifically detects Actin Alpha 1 Cardiac Muscle in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Actin Alpha 1 Cardiac Muscle
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 959615
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 1.7-25 ug/mL
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. P68032
Gene Alias ACTC, ACTC1, actin, alpha, cardiac muscle 1, ASD5, Cardiac alpha-Actin, CMD1R, CMH11, LVNC4
Host Species Mouse
Immunogen E. coli-derived recombinant human alpha-Cardiac Actin, Asp3-Phe377, Accession # P68032
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70
Test Specificity Detects human alpha-Cardiac Actin in direct ELISAs. Detects human,mouse, and rat alpha-Cardiac Actin in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.