Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat alpha-Cardiac Actin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $579.52
Specifications
| Antigen | Actin Alpha 1 Cardiac Muscle |
|---|---|
| Clone | 959606 |
| Dilution | Western Blot 0.1 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9308SP
![]() |
R&D Systems
MAB9308SP |
25 μg |
Each for $214.00
|
|
|||||
MAB9308100
![]() |
R&D Systems
MAB9308100 |
100 μg |
Each for $579.52
|
|
|||||
Description
Actin Alpha 1 Cardiac Muscle Monoclonal specifically detects Actin Alpha 1 Cardiac Muscle in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.Specifications
| Actin Alpha 1 Cardiac Muscle | |
| Western Blot 0.1 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat | |
| P68032 | |
| 70 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 959606 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ACTC, ACTC1, actin, alpha, cardiac muscle 1, ASD5, Cardiac alpha-Actin, CMD1R, CMH11, LVNC4 | |
| E.coli-derived recombinant human alpha-Cardiac Actin Asp3-Phe377 Accession # P68032 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human alpha-Cardiac Actin in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title