Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat AlphaB Crystallin/CRYAB Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4849SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4849SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4849SP Supplier R&D Systems Supplier No. MAB4849SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

AlphaB Crystallin/CRYAB Monoclonal specifically detects AlphaB Crystallin/CRYAB in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen AlphaB Crystallin/CRYAB
Applications Western Blot
Classification Monoclonal
Clone 731502
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Simple Western 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P02511
Gene Alias alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2, CRYA2alpha crystallin B chain, CRYAB, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
Gene Symbols CRYAB
Host Species Mouse
Immunogen E. coli-derived recombinant human Crystallin/CRYAB Met1-Lys175 Accession # P02511
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1410
Test Specificity Detects human AlphaB Crystallin/CRYAB in direct ELISAs and human, mouse, and rat AlphaB Crystallin/CRYAB in Western blots. In direct ELISAs, no cross-reactivity with recombinant human AlphaA Crystallin/CRYAA is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.