Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat AMPK beta 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2854
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2854 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2854 Supplier R&D Systems Supplier No. AF2854
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

AMPK beta 1 Polyclonal specifically detects AMPK beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen AMPK beta 1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q9Y478
Gene Alias 5'-AMP-activated protein kinase beta-1 subunit, AMP-activated protein kinase beta subunit, AMPK, AMPK beta -1 chain, AMPK beta 1,5'-AMP-activated protein kinase subunit beta-1, AMPK subunit beta-1, AMPKb, HAMPKb, MGC17785, PRKAB1, protein kinase, AMP-activated, beta 1 non-catalytic subunit, protein kinase, AMP-activated, noncatalytic, beta-1
Gene Symbols PRKAB1
Host Species Goat
Immunogen E. coli-derived recombinant human AMPK beta 1 Met1-Ile270 Accession # Q9Y478
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5564
Test Specificity Detects human, mouse, and rat AMPK beta 1 in Western blots. In Western blots, no cross-reactivity with recombinant human AMPK beta 2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.