Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Androgen R/NR3C4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5876
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5876 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5876 Supplier R&D Systems Supplier No. AF5876
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Androgen R/NR3C4 Polyclonal specifically detects Androgen R/NR3C4 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Androgen R/NR3C4
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. P10275
Gene Alias AIS, androgen receptor, AndrogenR, AR, DHTR, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SBMA, SMAX1, SMAX1SBMA, spinal and bulbar muscular atrophy
Gene Symbols AR
Host Species Sheep
Immunogen E. coli-derived recombinant human Androgen R/NR3C4 Thr660-Gln919 Accession # P10275
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367
Test Specificity Detects human Androgen R/NR3C4 in direct ELISAs. Detects human, mouse and rat Androgen R/NR3C4 in Western blotting.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.