Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Androgen R/NR3C4 APC-conjugated Antibody, R&D Systems™

Catalog No. IC5876A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5876A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5876A Supplier R&D Systems Supplier No. IC5876A
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Androgen R/NR3C4 Monoclonal specifically detects Androgen R/NR3C4 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Androgen R/NR3C4
Applications Flow Cytometry
Classification Monoclonal
Clone 523339
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P10275
Gene Alias AIS, androgen receptor, AndrogenR, AR, DHTR, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SBMA, SMAX1, SMAX1SBMA, spinal and bulbar muscular atrophy
Gene Symbols AR
Host Species Mouse
Immunogen E. coli-derived recombinant human Androgen R/NR3C4 Thr660-Gln919 Accession # P10275
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367
Test Specificity Detects human Androgen R/NR3C4 in direct ELISAs.
Target Species Human, Mouse, Rat
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.