Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Annexin A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB37701
This item is not returnable.
View return policy
Description
Annexin A1 Monoclonal specifically detects Annexin A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Knockout Validated.Specifications
Annexin A1 | |
Monoclonal | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL | |
P04083 | |
ANXA1 | |
E. coli-derived recombinant human Annexin A1 Met1-Asn346 Accession # P04083 | |
100 μg | |
Primary | |
Detects human Annexin A1 in direct ELISAs and human, mouse, and rat Annexin A1 in Western blots. In direct ELISAs, approximately 25%-50% cross-reactivity with recombinant human (rh) Annexin A2 is observed and no cross-reactivity with rhAnnexin A3, A6, or A11 is observed. In Western blots, no cross-reactivity with rhAnnexin A6 is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
686122 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, ANXA1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
301 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction