Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Annexin A2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3928
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3928 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3928 Supplier R&D Systems Supplier No. AF3928
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

Annexin A2 Polyclonal specifically detects Annexin A2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.

Specifications

Antigen Annexin A2
Applications Western Blot, Immunohistochemistry, KnockDown
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated
Gene Accession No. P07355
Gene Alias annexin A2, Annexin II, annexin-2, ANX2, ANX2L4, ANX2L4LPC2, ANXA2, CAL1H, Calpactin I heavy chain, calpactin I heavy polypeptide, Calpactin-1 heavy chain, chromobindin 8, chromobindin-8, LIP2, LIP2PAP-IV, Lipocortin II, Lipocortin-2, LPC2D, LPC2DP36, p36, PAP-IV, Placental anticoagulant protein IV, Protein I
Gene Symbols ANXA2
Host Species Goat
Immunogen E. coli-derived recombinant human Annexin A2 Met1-Asp339 Accession # P07355
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 302
Test Specificity Detects human, mouse, and rat Annexin A2 in Western blots. In Western blots, no cross-reactivity with recombinant human Annexin A1, A3, A4, A5, A6, A7, A8, A9, A10, A11, or A13 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.