Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat APE Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1044SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1044SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1044SP Supplier R&D Systems Supplier No. MAB1044SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

APE Monoclonal specifically detects APE in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen APE
Applications Western Blot
Classification Monoclonal
Clone 200913
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P27695
Gene Alias AP endonuclease 1, APE, APE1, APE-1, APEAPEX nuclease (multifunctional DNA repair enzyme), APENAP endonuclease class I, APEX deoxyribonuclease (apurinic or apyrimidinic), APEX nuclease, APEX nuclease (multifunctional DNA repair enzyme) 1, APEX1, Apurinic-apyrimidinic endonuclease 1, APXAP lyase, DNA-(apurinic or apyrimidinic site) lyase, EC 3.1, EC 4.2.99.18, HAP1apurinic/apyrimidinic (abasic) endonuclease, multifunctional DNA repair enzyme, protein REF-1, redox factor 1, Redox factor-1, REF-1, REF1 apurinic/apyrimidinic exonuclease
Gene Symbols APEX1
Host Species Mouse
Immunogen E. coli-derived recombinant human APE Pro2-Leu318 Accession # P27695
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 328
Test Specificity Detects human, mouse and rat APE in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.