Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat BAK Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF8161SP
This item is not returnable.
View return policy
Description
BAK Polyclonal specifically detects BAK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
BAK | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Apoptosis regulator BAK, BAK1, BAK-LIKE, BAKMGC3887, BCL2-antagonist/killer 1, Bcl2-L-7, BCL2L7, BCL2L7bcl-2 homologous antagonist/killer, BCL2-like 7 protein, Bcl-2-like protein 7, CDN1, MGC117255, pro-apoptotic protein BAK | |
Goat | |
Affinity Purified | |
RUO | |
578 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q16611 | |
BAK1 | |
E. coli-derived recombinant human BAK Pro20-Asn124 Accession # Q16611 | |
25 μg | |
Primary | |
Detects endogenous human, mouse and rat BAK in Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction