Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Bcl-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2889
This item is not returnable.
View return policy
Description
Bcl-10 Monoclonal specifically detects Bcl-10 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Bcl-10 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O95999 | |
BCL10 | |
E. coli-derived recombinant human Bcl-10 Met1-Leu192 Accession # O95999 | |
100 μg | |
Primary | |
Detects endogenous human, mouse, and rat Bcl-10 in Western blots. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
384202 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B-cell CLL/lymphoma 10CIPERCLAPB-cell lymphoma/leukemia 10, Bcl10, Bcl-10, CARD-containing apoptotic signaling protein, CARD-containing molecule enhancing NF-kappa-B, CARD-containing proapoptotic protein, CARD-like apoptotic protein, CARMEN, caspase-recruiting domain-containing protein, cCARMEN, c-E10, c-E10bcl-10, CED-3/ICH-1 prodomain homologous E10-like regulator, Cellular homolog of vCARMEN, cellular-E10, CIPER, CLAP, hCLAP, Mammalian CARD-containing adapter molecule E10, mE10, mE10CARD containing molecule enhancing NF-kB | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8915 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction