Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Bcl-xL Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $538.00
Specifications
| Antigen | Bcl-xL |
|---|---|
| Clone | 2052A |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 5 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Bcl-xL Monoclonal specifically detects Bcl-xL in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
| Bcl-xL | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BclxL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| E. coli-derived recombinant human Bcl-x L Met1-Arg212 Accession # Q07817 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2052A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q07817 | |
| 598 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Detects human Bcl-x L in direct ELISAs and detects human, mouse, and rat Bcl-x L in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title