Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat beta-2 Adrenergic R/ADRB2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB10040100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10040100 100 μg
MAB10040SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB10040100 Supplier R&D Systems Supplier No. MAB10040100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

beta-2 Adrenergic R/ADRB2 Monoclonal specifically detects beta-2 Adrenergic R/ADRB2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen beta-2 Adrenergic R/ADRB2
Applications Western Blot
Classification Monoclonal
Clone 2204C
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P07550
Gene Alias ADRB2, ADRB2R, ADRBR, adrenergic, beta-2-, receptor, surface, B2AR, B2ARADRB2R, BAR, beta-2 adrenergic receptor, beta2 AdrenergicR, beta-2 AdrenergicR, Beta-2 adrenoceptor, Beta-2 adrenoreceptor, BETA2AR, catecholamine receptor
Host Species Rabbit
Immunogen Synthetic peptide containing human beta-2 Adrenergic R/ADRB2, Accession No. P07550
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 154.0
Test Specificity Detects human beta-2 Adrenergic R/ADRB2 in direct ELISAs. Detects human, mouse, and rat beta-2 Adrenergic R/ADRB2 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.