Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat beta-Catenin Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC13292T100UG
This item is not returnable.
View return policy
Description
beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
beta-Catenin | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P35222 | |
CTNNB1 | |
E. coli-derived recombinant human beta-Catenin Ala2-Leu781 Accession # P35222 | |
RUO | |
1499 | |
Human, Mouse, Rat | |
IgG2a |
Flow Cytometry | |
196624 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
bCatenin, beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, CTNNB1, DKFZp686D02253, FLJ25606, FLJ37923 | |
Mouse | |
100 μg | |
Primary | |
Detects human, mouse, and rat beta-Catenin in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction