Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat BNIP3L Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4030
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4030 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4030 Supplier R&D Systems Supplier No. AF4030
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

BNIP3L Polyclonal specifically detects BNIP3L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen BNIP3L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. O60238
Gene Alias Adenovirus E1B19K-binding protein B5, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like, BCL2/adenovirus E1B 19-kd protein-interacting protein 3a, BCL2/adenovirus E1B 19kDa interacting protein 3-like, BNIP3a, BNIP3H, Nip3L, NIP3-like protein X, Nix, NIXBCL2/adenovirus E1B 19kD-interacting protein 3-like
Gene Symbols BNIP3L
Host Species Goat
Immunogen E. coli-derived recombinant human BNIP3L Ser2-Glu184 Accession # O60238
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 665
Test Specificity Detects endogenous human, mouse and rat BNIP3L in Western blots. In Western blots, less than 10% cross-reactivity to recombinant human BNIP3L is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.