Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat BTF3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5747
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5747 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5747 Supplier R&D Systems Supplier No. AF5747
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

BTF3 Polyclonal specifically detects BTF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen BTF3
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P20290
Gene Alias basic transcription factor 3, BETA-NAC, BTF3a, BTF3b, NACB, nascent-polypeptide-associated complex beta polypeptide, RNA polymerase B transcription factor 3, transcription factor BTF3
Gene Symbols BTF3
Host Species Goat
Immunogen E. coli-derived recombinant human BTF3 Lys46-Asn206 Accession # P20290
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 689
Test Specificity Detects human, mouse and rat BTF3 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.