Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Calcineurin A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2839
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2839 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2839 Supplier R&D Systems Supplier No. MAB2839
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Calcineurin A Monoclonal specifically detects Calcineurin A in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Calcineurin A
Applications Western Blot
Classification Monoclonal
Clone 302202
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNA, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1, CNA1CALNA1, EC 3.1.3.16, PP2B, PP3CA, PPP2B, PPP2BCCN1, PPP3CA, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Gene Symbols PPP3CA
Host Species Rat
Immunogen Bovine brain Calcineurin holoenzyme (A and B subunits)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5530
Test Specificity Detects human, mouse, and rat Calcineurin A.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.