Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat/Canine ALCAM/CD166 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 13 publications
Supplier: R&D Systems AF1172
This item is not returnable.
View return policy
Description
ALCAM/CD166 Polyclonal specifically detects ALCAM/CD166 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
ALCAM/CD166 | |
Polyclonal | |
Unconjugated | |
AAC06342 | |
ALCAM | |
Mouse myeloma cell line NS0-derived recombinant mouse ALCAM/CD166 Trp28-Lys527 Accession # AAC06342 | |
100 μg | |
Primary | |
Detects human, mouse and rat ALCAM/CD166 in Western blots. In direct ELISAs, less than 10% cross-reactivity with rhBCAM, recombinant mouse (rm) OCAM, and rmMAdCAM-1 is observed. Detects canine ALCAM/CD166 in flow cytometry. | |
Human, Mouse, Rat, Canine | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
LYOPH | |
Western Blot 0.2 ug/mL, Simple Western 12.5 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
Goat | |
Affinity Purified | |
RUO | |
214 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction