Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Carbonic Anhydrase III/CA3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF2185SP
This item is not returnable.
View return policy
Description
Carbonic Anhydrase III/CA3 Polyclonal specifically detects Carbonic Anhydrase III/CA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carbonic Anhydrase III/CA3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CA3, CAIII, CA-III, Car3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, EC 4.2.1.1 | |
Goat | |
Affinity Purified | |
RUO | |
761 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
P07451 | |
CA3 | |
E. coli-derived recombinant human Carbonic Anhydrase III/CA3 Ala2-Lys260 Accession # P07451 | |
25 μg | |
Primary | |
Detects human Carbonic Anhydrase III/CA3 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) CA1, rhCA2, rhCA4, rhCA9, rhCA10, rhCA13 and rhCA14 is observed. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction