Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Carboxylesterase 1/CES1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4920
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4920 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4920 Supplier R&D Systems Supplier No. AF4920
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Carboxylesterase 1/CES1 Polyclonal specifically detects Carboxylesterase 1/CES1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Carboxylesterase 1/CES1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2 ug/mL
Gene Accession No. NP_001020365
Gene Alias ACAT, acyl coenzyme A:cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, carboxylesterase 1, carboxylesterase 1 (monocyte/macrophage serine esterase 1), carboxylesterase 2 (liver), CEH, Ces-1, CES1, CES1A2, CES2EC 3.1.1.1, cholesteryl ester hydrolase, Cocaine carboxylesterase, EC 3.1.1, egasyn, HMSE1, HMSECES1A1, human monocyte/macrophage serine esterase 1, liver carboxylesterase 1, MGC117365, Monocyte/macrophage serine esterase, PCE-1, REH, Retinyl ester hydrolase, Serine esterase 1, Ses-1, SES1, SES1TGH, TGH, Triacylglycerol hydrolase
Gene Symbols CES1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase 1/CES1 His19-Glu563 Accession # NP_001020365
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1066
Test Specificity Detects human Carboxylesterase 1/CES1 in direct ELISAs and Western blots. Detects mouse Carboxylesterase 1/CES1 and rat Carboxylesterase 1/CES1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.