Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat CART Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF163SP
This item is not returnable.
View return policy
Description
CART/CARTPT Polyclonal specifically detects CART/CARTPT in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CART/CARTPT | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CART prepropeptide, CARTcocaine and amphetamine regulated transcript, CARTPT, cocaine- and amphetamine-regulated transcript protein | |
Goat | |
Affinity Purified | |
RUO | |
9607 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 1-15 ug/mL | |
Q16568 | |
CARTPT | |
E. coli-derived recombinant human CART Gln28-Leu116 Accession # Q16568 | |
25 μg | |
Primary | |
Detects human CART in direct ELISAs and Western blots. Also recognizes a truncated recombinant human CART containing amino acids 69-116. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction