Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Casein Kinase 1 alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4569SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4569SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4569SP Supplier R&D Systems Supplier No. AF4569SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Casein Kinase 1 alpha Polyclonal specifically detects Casein Kinase 1 alpha in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Casein Kinase 1 alpha
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001020276
Gene Alias casein kinase 1, alpha 1, casein kinase I isoform alpha, CK1CKI-alpha, CSNK1A1, down-regulated in lung cancer, EC 2.7.11.1, HLCDGP1, PRO2975
Gene Symbols CSNK1A1
Host Species Sheep
Immunogen E. coli-derived recombinant human CK1 alpha Ala255-Phe365 Accession # NP_001020276
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1452
Test Specificity Detects endogenous human, mouse, and rat CK1 alpha in Western blots. In Western blots, this antibody does not cross-react with recombinant human CK1 delta or CK1 gamma ; reactivity with CK1 epsilon is unknown.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.