Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat Caveolin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB5736
This item is not returnable.
View return policy
Description
Caveolin-1 Monoclonal specifically detects Caveolin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Caveolin-1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CAV, CAV1, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, Caveolin1, cell growth-inhibiting protein 32, MSTP085, VIP21 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
857 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunocytochemistry | |
7C8 | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P41350 | |
CAV1 | |
Puified rat adipocyte low density microsomes Accession # P41350 | |
100 μg | |
Primary | |
Detects endogenous human, mouse and rat Caveolin-1 in Western blots. | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction