Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Caveolin-1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM5736
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM5736 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM5736 Supplier R&D Systems Supplier No. BAM5736
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Caveolin-1 Monoclonal specifically detects Caveolin-1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry.

Specifications

Antigen Caveolin-1
Applications Immunocytochemistry
Classification Monoclonal
Clone 7C8
Conjugate Biotin
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and Sodium Azide with BSA as a carrier protein. with Sodium Azide
Gene Accession No. P41350
Gene Alias CAV, CAV1, caveolae protein, 22kD, caveolin 1, caveolae protein, 22kDa, Caveolin1, cell growth-inhibiting protein 32, MSTP085, VIP21
Gene Symbols CAV1
Host Species Mouse
Immunogen Purified rat adipocyte low density microsomes Accession # P41350
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 857
Test Specificity Caveolin-1 antibodies are ideal for immunocytochemistry colocalization studies in caveolae. The unconjugated antibody detects endogenous human, mouse and rat Caveolin-1 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.