Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Caveolin-2 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5788
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5788 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5788 Supplier R&D Systems Supplier No. BAF5788
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Caveolin-2 Polyclonal specifically detects Caveolin-2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry.

Specifications

Antigen Caveolin-2
Applications Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Immunocytochemistry 5-15 ug/mL
Gene Accession No. P51636
Gene Alias CAV, CAV2, caveolae protein, 20-kD, caveolin 2, caveolin 2 isoform a and b, caveolin-2, Caveolin2, MGC12294
Gene Symbols CAV2
Host Species Goat
Immunogen E. coli-derived recombinant human Caveolin-2 Met1-Lys86 Accession # P51636
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 858
Test Specificity Caveolin-2 antibodies are ideal for immunocytochemistry colocalization studies in caveolae. The unconjugated antibody detects endogenous human, mouse, and rat Caveolin-2 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.