Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat CDC25A Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB1648
This item is not returnable.
View return policy
Description
CDC25A Monoclonal specifically detects CDC25A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CDC25A | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
AAA58415 | |
CDC25A | |
E. coli-derived recombinant human CDC25A Glu2-Leu523 Accession # AAA58415 | |
100 μg | |
Primary | |
Detects human, mouse, and rat CDC25A in direct ELISAs and Western blots. In direct ELISA and Western blots, this antibody does not cross-react with recombinant human (rh) CDC25B or rhCDC25C. | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
336445 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CDC25A2, CDC25A2-CAG isoform, cell division cycle 25 homolog A (S. cerevisiae), cell division cycle 25 homolog A (S. pombe), cell division cycle 25A, Dual specificity phosphatase Cdc25A, EC 3.1.3.48, M-phase inducer phosphatase 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
993 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction