Learn More
Human/Mouse/Rat CDC25B Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CDC25B |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.3 ug/mL, Immunocytochemistry 10-25 ug/mL |
| Gene Accession No. | P30305 |
| Gene Alias | CDC25HU2, cell division cycle 25 homolog B (S. cerevisiae), cell division cycle 25 homolog B (S. pombe), cell division cycle 25B, Dual specificity phosphatase Cdc25B, EC 3.1.3.48, M-phase inducer phosphatase 2 |
| Gene Symbols | CDC25B |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.