Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat/Chicken Pax7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 28 publications
Supplier: R&D Systems MAB1675
This item is not returnable.
View return policy
Description
Pax7 Monoclonal specifically detects Pax7 in Human, Mouse, Rat, Chicken samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
Pax7 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
FLJ37460, HuP1, paired box 7, paired box gene 7, paired box homeotic gene 7, paired box protein Pax-7, paired domain gene 7, PAX7 transcriptional factor, PAX7B, RMS2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5081 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Immunocytochemistry | |
PAX7 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PAX7 | |
E. coli-derived recombinant chicken Pax7 aa 352-523 | |
100 μg | |
Primary | |
Detects human, mouse, rat, and chicken Pax7 (3-8). | |
Human, Mouse, Rat, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction