Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat Contactin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF904SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF904SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF904SP Supplier R&D Systems Supplier No. AF904SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 12 publications

Contactin-1 Polyclonal specifically detects Contactin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Neutralization.

Specifications

Antigen Contactin-1
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 1-5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. CAA79696
Gene Alias CNTN1, contactin 1, contactin-1, Contactin1, F11, F3, F3cam, Glycoprotein gp135, GP135, Neural cell surface protein F3
Gene Symbols CNTN1
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Contactin-1 Glu21-Ser993 Accession # CAA79696
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1272
Test Specificity Detects human Contactin-1 in direct ELISAs. Detects human, mouse, and rat Contactin-1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.